(2-Cyanoethyl)triethoxysilane, 97%, Thermo Scientific Chemicals
(2-Cyanoethyl)triethoxysilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2-Cyanoethyl)triethoxysilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L03543.14
also known as L03543-14
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.70 
Save 6.05 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.70 
Save 6.05 (15%)
Add to cart
(2-Cyanoethyl)triethoxysilane, 97%, Thermo Scientific Chemicals
Catalog numberL03543.14
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.70 
Save 6.05 (15%)
-
Add to cart
Chemical Identifiers
CAS919-31-3
IUPAC Name3-(triethoxysilyl)propanenitrile
Molecular FormulaC9H19NO3Si
InChI KeyGBQYMXVQHATSCC-UHFFFAOYSA-N
SMILESCCO[Si](CCC#N)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Frank Bauer; Hans-Jürgen Gläsel; Ulrich Decker; Horst Ernst; Annette Freyer; Eberhard Hartmann; Volker Sauerland; Reiner Mehnert. Trialkoxysilane grafting onto nanoparticles for the preparation of clear coat polyacrylate systems with excellent scratch performance. Progress in Organic Coatings. 2003, 47, (2), 147-153.
  2. Daniel A. Higgins; Maryanne M. Collinson; Ginagunta Saroja and Angela M. Bardo. Single-Molecule Spectroscopic Studies of Nanoscale Heterogeneity in Organically Modified Silicate Thin Films. Chem. Mater. 2002, 14, (9), 3734-3744.