(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only
General References:
- Frank Bauer; Hans-Jürgen Gläsel; Ulrich Decker; Horst Ernst; Annette Freyer; Eberhard Hartmann; Volker Sauerland; Reiner Mehnert. Trialkoxysilane grafting onto nanoparticles for the preparation of clear coat polyacrylate systems with excellent scratch performance. Progress in Organic Coatings. 2003, 47, (2), 147-153.
- Daniel A. Higgins; Maryanne M. Collinson; Ginagunta Saroja and Angela M. Bardo. Single-Molecule Spectroscopic Studies of Nanoscale Heterogeneity in Organically Modified Silicate Thin Films. Chem. Mater. 2002, 14, (9), 3734-3744.