Thermo Scientific Chemicals

N-Cyclohexylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L03569.06
5 g, Each
Thermo Scientific Chemicals

N-Cyclohexylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L03569.06
5 g, Each
Quantity
Catalog number: L03569.06
also known as L03569-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25100-12-3
IUPAC Name
hydrogen N-cyclohexylhydroxylamine chloride
Molecular Formula
C6H14ClNO
InChI Key
SSVAHXZUFFSFER-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ONC1CCCCC1
Form
Crystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)
White to cream to pale brown or pink
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Cyclohexylhydroxylamine hydrochloride is used in the preparation of 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone and hexahydroisoxazolequinoline analogs. It is also used in the preparation of N-cyclohexyl nitrone by reaction with aldehyde. Further, it acts as a hydroxylamination reagent for allyl esters. In addition to this, it serves as a reagent in the conversion of 1,2-dicarboxylic acids to alfa-beta-unsaturated carboxylic acids.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text