2,4-Dimethylpentane, 98+%, Thermo Scientific Chemicals
2,4-Dimethylpentane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dimethylpentane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number L03570.09
also known as L03570-09
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
Add to cart
2,4-Dimethylpentane, 98+%, Thermo Scientific Chemicals
Catalog numberL03570.09
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-08-7
IUPAC Name2,4-dimethylpentane
Molecular FormulaC7H16
InChI KeyBZHMBWZPUJHVEE-UHFFFAOYSA-N
SMILESCC(C)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)>98.0%
Refractive Index1.3800-1.3840 @ 20?C
It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

Solubility
Soluble in acetone, benzene, chloroform, alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Dennis D. Tanner; Parviz M. Rahimi. Free-radical cyanation. Novel free-radical reaction of methyl cyanoformate or cyanogen with 2,4-dimethylpentane. J. Org. Chem. 1979, 44, (10), 1674-1677.
  2. H. Moses Chung; Clifford C. Walters; Steven Buck; Gail Bingham. Mixed signals of the source and thermal maturity for petroleum accumulations from light hydrocarbons: an example of the Beryl field. Organic Geochemistry. 1998, 29, (1-3), 381-396.