Thermo Scientific Chemicals

1-Butylamine, 99%, Thermo Scientific Chemicals

Catalog number: L03575.AP
500 mL, Each
Thermo Scientific Chemicals

1-Butylamine, 99%, Thermo Scientific Chemicals

Catalog number: L03575.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03575.AP
also known as L03575-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-73-9
IUPAC Name
butan-1-amine
Molecular Formula
C4H11N
InChI Key
HQABUPZFAYXKJW-UHFFFAOYSA-N
SMILES
CCCCN
Form
Liquid
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.3990-1.4030 @ 20?C

Description

1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N'-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N′-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.

Solubility
Miscible with ethanol and water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, aluminum, copper, copper alloys and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text