Thermo Scientific Chemicals

Cyclopentanethiol, 97%, Thermo Scientific Chemicals

Catalog number: L03577.06
5 g, Each
Thermo Scientific Chemicals

Cyclopentanethiol, 97%, Thermo Scientific Chemicals

Catalog number: L03577.06
5 g, Each
Quantity
Catalog number: L03577.06
also known as L03577-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1679-07-8
IUPAC Name
cyclopentanethiol
Molecular Formula
C5H10S
InChI Key
WVDYBOADDMMFIY-UHFFFAOYSA-N
SMILES
SC1CCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
>96.0%
Refractive Index
1.4875-1.4925 @ 20?C

Description

Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanethiol has used as a food spice. It is a pharmaceutical intermediate.

Solubility
Slightly soluble in water; soluble in alcohol and oils.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text