Thermo Scientific Chemicals

Neopentyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: L03579.06
5 g, Each
Thermo Scientific Chemicals

Neopentyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: L03579.06
5 g, Each
Quantity
Catalog number: L03579.06
also known as L03579-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
630-17-1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Infrared spectrum
Conforms (U.S. specification)
Refractive Index
1.4330-1.4390 @ 20°C (non-U.S. specification)

Description

Neopentyl bromide is used in synthesis of S-alkylated cysteine derivatives with branched alkyl chains. It can be used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neopentyl bromide is used in synthesis of S-alkylated cysteine derivatives with branched alkyl chains. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in ethanol, ether, aetone and benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text