Thermo Scientific Chemicals

Chlorodiphenylsilane, tech. 90%, Thermo Scientific Chemicals

Catalog number: L03602.06
5 g, Each
Thermo Scientific Chemicals

Chlorodiphenylsilane, tech. 90%, Thermo Scientific Chemicals

Catalog number: L03602.06
5 g, Each
Quantity
Catalog number: L03602.06
also known as L03602-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1631-83-0
IUPAC Name
chlorodiphenylsilyl
Molecular Formula
C12H10ClSi
InChI Key
YCITZMJNBYYMJO-UHFFFAOYSA-N
SMILES
Cl[Si](C1=CC=CC=C1)C1=CC=CC=C1
Form
Liquid
Assay (GC)
≥88.0%
Appearance (Color)
Clear, colorless to pale yellow
Identification (FTIR)
Conforms
Refractive Index
1.5770-1.5850 @ 20?C

Description

Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text