Thermo Scientific Chemicals

(+/-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals

Catalog number: L03608.14
25 g, Each
Thermo Scientific Chemicals

(+/-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals

Catalog number: L03608.14
25 g, Each
Quantity
Catalog number: L03608.14
also known as L03608-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6168-72-5
IUPAC Name
2-aminopropan-1-ol
Molecular Formula
C3H9NO
InChI Key
BKMMTJMQCTUHRP-UHFFFAOYNA-N
SMILES
CC(N)CO
Appearance (Color)
Clear colorless to yellow
Form
Liquid or viscous liquid
Refractive Index
1.4485-1.4525 @ 20°C
Assay (GC)
≥95.0%

Description

DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.

Solubility
Fully miscible in water.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text