Thermo Scientific Chemicals

Triphenylarsine, 97%, Thermo Scientific Chemicals

Catalog number: L03616.06
5 g, Each
Thermo Scientific Chemicals

Triphenylarsine, 97%, Thermo Scientific Chemicals

Catalog number: L03616.06
5 g, Each
Quantity
Catalog number: L03616.06
also known as L03616-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
603-32-7
IUPAC Name
triphenylarsane
Molecular Formula
C18H15As
InChI Key
BPLUKJNHPBNVQL-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)[As](C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms (non-U.S. specification)
Melting Point
56.0-65.0°C (non-U.S. specification)
Appearance (Color)
White to pale yellow

Description

Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Handle and store under inert gas. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text