2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Soluble in water (24 mg/ml at 25°C).
Notes
Storage temperature: 2 - 4°C. Incompatible with Strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
D.L. Capone; K.A. Van Leeuwen; K.H. Pardon; M.A. Daniel; G.M. Elsey; A.D. Coulter And M.A. Sefton. Identification and analysis of 2-chloro-6-methylphenol, 2,6-dichlorophenol and indole: causes of taints and off-flavours in wines.Australian Journal of Grape and Wine Research.2010, 16 210-217.