Thermo Scientific Chemicals

exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: L03620.22
100 g, Each
Thermo Scientific Chemicals

exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: L03620.22
100 g, Each
Quantity
Catalog number: L03620.22
also known as L03620-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6118-51-0
IUPAC Name
4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Formula
C8H6O4
InChI Key
QQYNRBAAQFZCLF-UHFFFAOYNA-N
SMILES
O=C1OC(=O)C2C3OC(C=C3)C12
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%

Description

Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text