Thermo Scientific Chemicals

Ethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L03624.0B
1000 g, Each
Thermo Scientific Chemicals

Ethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L03624.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03624.0B
also known as L03624-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-66-4
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Appearance (Color)
Colorless to white to cream
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤2%

Description

Ethylaminehydrochloride is used as a raw material for organic synthesis. Its free base ethylamine is used in the diethyldiazene, dimethylolethyltriazone, ethylcyanopyrolidone disperse and 1,3-diethylthiourea. It is a precursor used for the preparation of benzilnitrate, detergents, rayon, rocket propellant and alkyl isocyanates, which finds application in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text