Thermo Scientific Chemicals

2-Methyl-1-phenyl-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog number: L03679.22
100 g, Each
Thermo Scientific Chemicals

2-Methyl-1-phenyl-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog number: L03679.22
100 g, Each
Quantity
Catalog number: L03679.22
also known as L03679-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-86-7
IUPAC Name
2-methyl-1-phenylpropan-2-ol
Molecular Formula
C10H14O
InChI Key
RIWRBSMFKVOJMN-UHFFFAOYSA-N
SMILES
CC(C)(O)CC1=CC=CC=C1
Appearance (Color)
Colorless to white to pale yellow
Form
Crystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Assay (GC)
≥98.0%
Refractive Index
1.5135-1.5175 @ 20?C
Identification (FTIR)
Conforms

Description

2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text