Chemical Identifiers
CAS362-43-6
IUPAC Name1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC12H16N2O6
InChI KeyGFDUSNQQMOENLR-PEBGCTIMSA-N
SMILESCC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=CC(=O)NC1=O
View more