Thermo Scientific Chemicals

Cyclohexyl methyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: L03712.14
25 g, Each
Thermo Scientific Chemicals

Cyclohexyl methyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: L03712.14
25 g, Each
Quantity
Catalog number: L03712.14
also known as L03712-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7133-37-1
IUPAC Name
(methylsulfanyl)cyclohexane
Molecular Formula
C7H14S
InChI Key
QQBIOCGHCKNYGP-UHFFFAOYSA-N
SMILES
CSC1CCCCC1
Refractive Index
1.4930-1.4980 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%

Description

Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in water 173.5 mg/L @ 25°C, alcohol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text