Methyl 4-chlorobutyrate is used as a pharmaceutical synthesis, industrial dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-chlorobutyrate is used as a pharmaceutical synthesis, industrial dye intermediates.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Thomas H. Johnson; Thomas F. Baldwin. Interaction of cyclopropane with platinum-metal chlorides under carbon monoxide pressure in benzene. J. Org. Chem. 1980, 45, (1), 140-142.
Martin Himly, PhD.; Beatrice Jahn-Schmid, PhD.; Klaus Pittertschatscher, PhD.; Barbara Bohle, PhD.; Karl Grubmayr, PhD.; Fátima Ferreira, PhD.; Herwig Ebner, MD.; Christof Ebner, MD. IgE-mediated immediate-type hypersensitivity to the pyrazolone drug propyphenazone. Journal of Allergy and Clinical Immunology. 2003, 111, (4), 882-888.