Thermo Scientific Chemicals

4,4'-Diaminoazobenzene, 95%, Thermo Scientific Chemicals

Catalog number: L03743.MD
250 mg, Each
Thermo Scientific Chemicals

4,4'-Diaminoazobenzene, 95%, Thermo Scientific Chemicals

Catalog number: L03743.MD
250 mg, Each
Quantity
Catalog number: L03743.MD
also known as L03743-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
538-41-0
Form
Crystals or powder or crystalline powder or needles
Assay (HPLC)
≥94.0%
Appearance (Color)
Yellow to orange to dark brown to brown

Description

4,4'-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.

Solubility
Very soluble in ethanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text