Thermo Scientific Chemicals

Cyanamide, 98+%, stab., Thermo Scientific Chemicals

Catalog number: L03744.22
100 g, Each
Thermo Scientific Chemicals

Cyanamide, 98+%, stab., Thermo Scientific Chemicals

Catalog number: L03744.22
100 g, Each
Quantity
Catalog number: L03744.22
also known as L03744-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
420-04-2
IUPAC Name
aminoformonitrile
Molecular Formula
CH2N2
InChI Key
XZMCDFZZKTWFGF-UHFFFAOYSA-N
SMILES
NC#N
Appearance (Color)
White
Form
Crystalline powder or crystals
Assay (unspecified)
≥98.0%
Impurity content
≤0.3% Dicyandiamide
Water Content (Karl Fischer Titration)
≤1.0%

Description

Cyanamide is used as an intermediate in the production of pharmaceuticals and other organic compounds. It is also used as an alcohol deterrent drug. It is widely used as plant growth regulator, agrochemical and fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanamide is used as an intermediate in the production of pharmaceuticals and other organic compounds. It is also used as an alcohol deterrent drug. It is widely used as plant growth regulator, agrochemical and fertilizer.

Solubility
Soluble in water, butanol, methyl ethyl ketone, ethyl acetate, alcohols, phenols, chloroform, amines and ethers. Slightly soluble in carbon disufide. Insoluble in cyclohexane.

Notes
Incompatible with barium peroxide, boric acid, dry hydrogen and hydrogen peroxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text