Thermo Scientific Chemicals

Methyl phenylsulfonylacetate, 98%, Thermo Scientific Chemicals

Catalog number: L03748.14
25 g, Each
Thermo Scientific Chemicals

Methyl phenylsulfonylacetate, 98%, Thermo Scientific Chemicals

Catalog number: L03748.14
25 g, Each
Quantity
Catalog number: L03748.14
also known as L03748-14
Price (USD)
Quantity
-
Have Questions?

Description

Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text