L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.
Solubility
Soluble in water, methanol (50 mg/ml).
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture, heat.
RUO – Research Use Only
General References:
- Stone T. W.Cortical pyramidal tract interneurones and their sensitivity to L-glutamic acid. The Journal of Physiology. 1973, 233, (1), 211-225.
- James S. Nowick; Noel A. Powell; Tram M. Nguyen; Glenn Noronha. An improved method for the synthesis of enantiomerically pure amino acid ester isocyanates. J. Org. Chem. 1992, 57, (26), 7364-7366.