Thermo Scientific Chemicals

beta-Estradiol, 99% (dry wt.), ca 3% water, Thermo Scientific Chemicals

Catalog number: L03801.06
5 g, Each
Thermo Scientific Chemicals

beta-Estradiol, 99% (dry wt.), ca 3% water, Thermo Scientific Chemicals

Catalog number: L03801.06
5 g, Each
Quantity
Catalog number: L03801.06
also known as L03801-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-28-2
IUPAC Name
(1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Molecular Formula
C18H24O2
InChI Key
VOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILES
C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O
Form
Powder
Assay (Silylated GC)
> 98.5%
Water Content (Karl Fischer Titration)
<4.0%
Optical Rotation
+79.0? ? 3.0? (c=1 in Dioxane)
Appearance (Color)
White

Description

β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner's syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner′s syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

Solubility
Soluble in dimethyl sulfoxide, ethanol , water, phosphate buffer saline, dimethyl formamide, acetone, dioxane and alkali hydroxides. Slightly soluble in vegetable oils.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text