Berberine chloride hydrate, 96%, water <17%, Thermo Scientific Chemicals
Berberine chloride hydrate, 96%, water &lt;17%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Berberine chloride hydrate, 96%, water <17%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L03807.14
also known as L03807-14
Price (USD)/ Each
95.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
95.50
Add to cart
Berberine chloride hydrate, 96%, water <17%, Thermo Scientific Chemicals
Catalog numberL03807.14
Price (USD)/ Each
95.50
-
Add to cart
Chemical Identifiers
CAS141433-60-5
IUPAC Name16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-13-ylium chloride
Molecular FormulaC20H18ClNO4
InChI KeyVKJGBAJNNALVAV-UHFFFAOYSA-M
SMILES[Cl-].COC1=C(OC)C2=C[N+]3=C(C=C2C=C1)C1=CC2=C(OCO2)C=C1CC3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Water Content (Karl Fischer Titration)≥5.0 to ≤17.0%
Assay (Titration ex Chloride)≥95.0% to ≤105.0% (dry weight basis)
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

Solubility
Soluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
RUO – Research Use Only

General References:

  1. Xiaoqing Wang; Shijing Qiu; Xiao Yao; Tingting Tang; Kerong Dai and Zhen'an Zhu. Berberine inhibits Staphylococcus Epidermidis adhesion and biofilm formation on the surface of titanium alloy. Journal of Orthopaedic Research. 2009, 27, (11), 1487-1492.
  2. Xiaoqing Wang; Xiao Yao; Zhen’an Zhu; Tingting Tang; Kerong Dai; Irina Sadovskaya; Sigrid Flahaut; Said Jabbouri. Effect of berberine on Staphylococcus epidermidis biofilm formation. International Journal of Antimicrobial Agents. 2009, 34, (1), 60-66.