Thermo Scientific Chemicals

Methyl 2-hydroxy-4-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L03855.09
10 g, Each
Thermo Scientific Chemicals

Methyl 2-hydroxy-4-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L03855.09
10 g, Each
Quantity
Catalog number: L03855.09
also known as L03855-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5446-02-6
IUPAC Name
methyl 2-hydroxy-4-methoxybenzoate
Molecular Formula
C9H10O4
InChI Key
ZICRWXFGZCVTBZ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(OC)C=C1O
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
47.0-53.0?C

Description

Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text