16-Hexadecanolide, 97%, Thermo Scientific Chemicals
16-Hexadecanolide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

16-Hexadecanolide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L03864.18
also known as L03864-18
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
Add to cart
16-Hexadecanolide, 97%, Thermo Scientific Chemicals
Catalog numberL03864.18
Price (USD)/ Each
232.65
Online exclusive
258.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS109-29-5
IUPAC Name1-oxacycloheptadecan-2-one
Molecular FormulaC16H30O2
InChI KeyLOKPJYNMYCVCRM-UHFFFAOYSA-N
SMILESO=C1CCCCCCCCCCCCCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Appearance (Color)Colorless to white to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Melting Point (clear melt)30.0-39.0?C
16-Hexadecanolide is used in the manfacturing of perfumes. It acts as masking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
16-Hexadecanolide is used in the manfacturing of perfumes. It acts as masking agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Shuhei Namekawa; Hiroshi Uyama; Shiro Kobayashi. Lipase-catalyzed ring-opening polymerization of 16-hexadecanolide..Proceedings of the Japan Academy.1998, 74 65-68.
  2. V. N. Emel’yanenko; S. P. Verevkin; E. N. Stepurko; G. N. Rogano. The thermodynamic characteristics of 15-pentadecanolide and 16-hexadecanolide..Russian Journal of Physical Chemistry A.2011, 85 348-356.