Thermo Scientific Chemicals

Diallyl maleate, 95%, Thermo Scientific Chemicals

Catalog number: L03867.0B
1000 g, Each
Thermo Scientific Chemicals

Diallyl maleate, 95%, Thermo Scientific Chemicals

Catalog number: L03867.0B
1000 g, Each
Quantity
Catalog number: L03867.0B
also known as L03867-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
999-21-3
IUPAC Name
1,4-bis(prop-2-en-1-yl) (2Z)-but-2-enedioate
Molecular Formula
C10H12O4
InChI Key
ZPOLOEWJWXZUSP-WAYWQWQTSA-N
SMILES
C=CCOC(=O)C=C/C(=O)OCC=C
Form
Liquid
Free acid (titration)
≤2.5% (non-U.S. specification)
Refractive Index
1.4660-1.4720 @ 20°C (non-U.S. specification)
Infrared spectrum
Conforms (U.S. specification)
Appearance (Color)
Clear colorless to yellow

Description

Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.

Solubility
Soluble in water 151.2 mg/L @ 25°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text