7-Methyl-8-nitroquinoline is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
7-Methyl-8-nitroquinoline is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Robert C. Elderfield; Walter J. Gensler; Thurmond A. Williamson; John M. Griffing; S. Morris Kupchan; John T. Maynard; Frank J. Kreysa; John B. Wright. Synthesis of 5-Substituted Derivatives of 6-Methoxy-8-aminoquinoline and of 5-Chloro-6-methoxyquinoline. J. Am. Chem. Soc. 1946, 68, (8), 1584-1587.
J. Curtis Shoup; John A. BurkeJr. Kinetics and mechanism of the reactions of substituted 8-mercaptoquinoline complexes of nickel(II) with some alkyl halides. Inorg. Chem. 1973, 12, (8), 1851-1855.