Thermo Scientific™

(R)-(-)-2-Aminobutane, 99%, Thermo Scientific Chemicals

Catalog number: L03889.MD
250 mg, Each
Thermo Scientific™

(R)-(-)-2-Aminobutane, 99%, Thermo Scientific Chemicals

Catalog number: L03889.MD
250 mg, Each
Quantity
Catalog number: L03889.MD
also known as L03889-MD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13250-12-9
IUPAC Name
butan-2-amine
Molecular Formula
C4H11N
InChI Key
BHRZNVHARXXAHW-UHFFFAOYNA-N
SMILES
CCC(C)N
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Optical Rotation
-8.0? ? 0.5? (neat)

Description

(R)-(-)-2-Aminobutane is a chiral amine plays an important role in pharmaceutical chemistry and in organic synthesis as chiral auxiliary, which is used to determine the enantiomeric excess (ee).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-2-Aminobutane is a chiral amine plays an important role in pharmaceutical chemistry and in organic synthesis as chiral auxiliary, which is used to determine the enantiomeric excess (ee).

Solubility
Miscible with water.

Notes
Air and light sensitive. Store it in a cool place. Incompatible with acids, acid chloride, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text