Thermo Scientific Chemicals

3-Methylindole, 99%, Thermo Scientific Chemicals

Catalog number: L03890.06
5 g, Each
Thermo Scientific Chemicals

3-Methylindole, 99%, Thermo Scientific Chemicals

Catalog number: L03890.06
5 g, Each
Quantity
Catalog number: L03890.06
also known as L03890-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-34-1
IUPAC Name
3-methyl-1H-indole
Molecular Formula
C9H9N
InChI Key
ZFRKQXVRDFCRJG-UHFFFAOYSA-N
SMILES
CC1=CNC2=CC=CC=C12
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
94-98?C
Appearance (Color)
White to pale brown
Form
Crystals or powder or crystalline powder or flakes

Description

3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

Solubility
Soluble in water, Ether, Alcohols, Benzene, Acetone, Chloroform.

Notes
Light Sensitive. Store in dark away from oxidizing agents. Incompatible with light, strong acids, acid ahydrides, acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text