L-(+)-Norleucine, 99%, Thermo Scientific Chemicals
L-(+)-Norleucine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-(+)-Norleucine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L03913.06
also known as L03913-06
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
Add to cart
L-(+)-Norleucine, 99%, Thermo Scientific Chemicals
Catalog numberL03913.06
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS327-57-1
IUPAC Name(2S)-2-aminohexanoic acid
Molecular FormulaC6H13NO2
InChI KeyLRQKBLKVPFOOQJ-YFKPBYRVSA-N
SMILESCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation+23.0 ± 1.0? (c=5 in 5N HCl)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only