Acetophenone azine, 97%, Thermo Scientific Chemicals
Acetophenone azine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetophenone azine, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Catalog number L03924.06
also known as L03924-06
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
Add to cart
Acetophenone azine, 97%, Thermo Scientific Chemicals
Catalog numberL03924.06
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS729-43-1
IUPAC Name(E,E)-bis(1-phenylethylidene)hydrazine
Molecular FormulaC16H16N2
InChI KeyMOKMQSIJAHPSQX-HBKJEHTGSA-N
SMILESCC(=N/N=C(C)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or crystalline powder and/or powder
Assay (GC)≥96.0%
Melting Point (clear melt)120-124?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only