Thermo Scientific Chemicals

Ethyl phenylcyanoacetate, 98%, Thermo Scientific Chemicals

Catalog number: L03929.14
25 g, Each
Thermo Scientific Chemicals

Ethyl phenylcyanoacetate, 98%, Thermo Scientific Chemicals

Catalog number: L03929.14
25 g, Each
Quantity
Catalog number: L03929.14
also known as L03929-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4553-07-5
IUPAC Name
ethyl 2-cyano-2-phenylacetate
Molecular Formula
C11H11NO2
InChI Key
SXIRJEDGTAKGKU-UHFFFAOYNA-N
SMILES
CCOC(=O)C(C#N)C1=CC=CC=C1
Assay (GC)
≥97.5%
Refractive Index
1.5025-1.5075 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text