5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals
5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L03952.14
also known as L03952-14
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
Add to cart
5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals
Catalog numberL03952.14
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
CAS5414-21-1
IUPAC Name5-bromopentanenitrile
Molecular FormulaC5H8BrN
InChI KeyNWWWGAKVHCSAEU-UHFFFAOYSA-N
SMILESBrCCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥98.0%
Refractive Index1.4765-1.4805 @ 20?C
5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

Solubility
Slightly soluble in water (4.1 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. David Griller.; Peter Schmid.; Keith U. Ingold. Cyclization of the 4-cyanobutyl radical. Canadian Journal of Chemistry. 1979, 57 (7), 831-834.
  2. Xiaoyan Shu.; Wei Chen.; Dabin Hou.; Qingbin Meng.; Renlin Zheng.; Chunju Li. Novel binding regioselectivity in the interpenetration of a non-symmetric axle into a non-symmetric pillar[5]arene wheel. Chem. Commun. 2014, 50 (37), 4820-4823.