Thermo Scientific Chemicals

2,3-Dimercaptopropanol, 97%, Thermo Scientific Chemicals

Catalog number: L03953.06
5 g, Each
Thermo Scientific Chemicals

2,3-Dimercaptopropanol, 97%, Thermo Scientific Chemicals

Catalog number: L03953.06
5 g, Each
Quantity
Catalog number: L03953.06
also known as L03953-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-52-9
IUPAC Name
2,3-disulfanylpropan-1-ol
Molecular Formula
C3H8OS2
InChI Key
WQABCVAJNWAXTE-UHFFFAOYNA-N
SMILES
OCC(S)CS
Appearance (Color)
Clear colorless to pale yellow
Assay (Iodometric Titration)
≥96.0% to ≤104.0%
Form
Liquid
Refractive Index
1.5700-1.5750 @ 20°C

Description

British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.2,3-Dimercaptopropanol is used as a chelating agent involved in the treatment of arsenic, antimony, lead, gold and mercury poisoning. It is also used as an antidote to the warfare agent Lewisite. Further, it is used for the treatment of Wilson's disease. In addition to this, it is used as an adjunct in the treatment of the acute encephalopathy of lead toxicity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ether, ethanol, vegetable oils and acetone. Slightly miscible with chloroform.

Notes
Air sensitive. Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text