Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 20% aq. soln., Thermo Scientific Chemicals

Catalog number: L03968.36
500 g, Each
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 20% aq. soln., Thermo Scientific Chemicals

Catalog number: L03968.36
500 g, Each
Quantity
Catalog number: L03968.36
also known as L03968-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-98-5
IUPAC Name
tetraethylazanium hydroxide
Molecular Formula
C8H21NO
InChI Key
LRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES
[OH-].CC[N+](CC)(CC)CC
Appearance (Color)
Clear, colorless
Form
Liquid
Assay from Suppliers CofA
18.0 - 22.0%

Description

Tetraethylammonium hydroxide is used as structure Directing Agents (SDAs), in selective etching, photovoltaic chemicals, advanced ceramics, strong organic base, flat panel display chemicals and transesterification catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium hydroxide is used as structure Directing Agents (SDAs), in selective etching, photovoltaic chemicals, advanced ceramics, strong organic base, flat panel display chemicals and transesterification catalyst.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text