Thermo Scientific Chemicals

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L03987.06
5 g, Each
Thermo Scientific Chemicals

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L03987.06
5 g, Each
Quantity
Catalog number: L03987.06
also known as L03987-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3588-17-8
IUPAC Name
(2E,4E)-hexa-2,4-dienedioic acid
Molecular Formula
C6H6O4
InChI Key
TXXHDPDFNKHHGW-ZPUQHVIOSA-N
SMILES
OC(=O)C=CC=CC(O)=O
Identification (FTIR)
Conforms
Form
Powder
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White to cream to pale brown
Assay (Titration)
≥97.5 to ≤102.5%

Description

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text