Thermo Scientific Chemicals

1,4-Butanesultone, 99%, Thermo Scientific Chemicals

Catalog number: L03992.22
100 g, Each
Thermo Scientific Chemicals

1,4-Butanesultone, 99%, Thermo Scientific Chemicals

Catalog number: L03992.22
100 g, Each
Quantity
Catalog number: L03992.22
also known as L03992-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1633-83-6
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4625-1.4655 @ 20?C

Description

1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.

Solubility
Miscible with chloroform and methanol. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text