Thermo Scientific Chemicals

4-Benzoylbutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: L04013.14
25 g, Each
Thermo Scientific Chemicals

4-Benzoylbutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: L04013.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L04013.14
also known as L04013-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1501-05-9
IUPAC Name
5-oxo-5-phenylpentanoic acid
Molecular Formula
C11H12O3
InChI Key
SHKWSBAVRQZYLE-UHFFFAOYSA-N
SMILES
OC(=O)CCCC(=O)C1=CC=CC=C1
Appearance (Color)
White to cream to yellow or pale briown
Form
Powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
122.0-132.0°C

Description

4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text