Thermo Scientific Chemicals

Ethyl hydrogen pimelate, 98%, Thermo Scientific Chemicals

Catalog number: L04025.06
5 g, Each
Thermo Scientific Chemicals

Ethyl hydrogen pimelate, 98%, Thermo Scientific Chemicals

Catalog number: L04025.06
5 g, Each
Quantity
Catalog number: L04025.06
also known as L04025-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33018-91-6
IUPAC Name
7-ethoxy-7-oxoheptanoic acid
Molecular Formula
C9H16O4
InChI Key
NQYXFXWKKYGBNL-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCCCC(O)=O
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Refractive Index
1.4400-1.4440 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text