Thermo Scientific Chemicals

Methyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L04059.AK
250 mL, Each
Thermo Scientific Chemicals

Methyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L04059.AK
250 mL, Each
Quantity
Catalog number: L04059.AK
also known as L04059-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2031-67-6
IUPAC Name
triethoxy(methyl)silane
Molecular Formula
C7H18O3Si
InChI Key
CPUDPFPXCZDNGI-UHFFFAOYSA-N
SMILES
CCO[Si](C)(OCC)OCC
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.3810-1.3860 @ 20?C
Assay (GC)
≥97.5%

Description

Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text