Thermo Scientific Chemicals

1,3-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: L04110.14
25 g, Each
Thermo Scientific Chemicals

1,3-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: L04110.14
25 g, Each
Quantity
Catalog number: L04110.14
also known as L04110-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
504-01-8
IUPAC Name
cyclohexane-1,3-diol
Molecular Formula
C6H12O2
InChI Key
RLMGYIOTPQVQJR-UHFFFAOYNA-N
SMILES
OC1CCCC(O)C1
Appearance (Color)
Colorless to white
Form
Crystals or powder or crystalline powder or fused solid or waxy solid or clear liquid or viscous liquid as melt
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥97.5%

Description

1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexanediol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text