Thermo Scientific Chemicals

1-Octadecanethiol, 96%, Thermo Scientific Chemicals

Catalog number: L04123.14
25 g, Each
Thermo Scientific Chemicals

1-Octadecanethiol, 96%, Thermo Scientific Chemicals

Catalog number: L04123.14
25 g, Each
Quantity
Catalog number: L04123.14
also known as L04123-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2885-00-9
IUPAC Name
octadecane-1-thiol
Molecular Formula
C18H38S
InChI Key
QJAOYSPHSNGHNC-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCS
Form
Fused solid or powder and/or lumps
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Appearance (Color)
White
Melting Point (clear melt)
29-34?C

Description

1-Octadecanethiol (ODT) is used to deposit thiolated molecules onto the surface of gold substrates, such as SAMs on gold quartz crystal resonators, on the tip of atomic force microscope probes, capacitive sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecanethiol (ODT) is used to deposit thiolated molecules onto the surface of gold substrates, such as SAMs on gold quartz crystal resonators, on the tip of atomic force microscope probes, capacitive sensors.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text