(Ethylthio)acetone, 95%, Thermo Scientific Chemicals
(Ethylthio)acetone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Ethylthio)acetone, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L04142.14
also known as L04142-14
Price (USD)/ Each
151.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
151.00
Add to cart
(Ethylthio)acetone, 95%, Thermo Scientific Chemicals
Catalog numberL04142.14
Price (USD)/ Each
151.00
-
Add to cart
Chemical Identifiers
CAS20996-62-7
IUPAC Name1-(ethylsulfanyl)propan-2-one
Molecular FormulaC5H10OS
InChI KeyZKDXKBIOEVBFGV-UHFFFAOYSA-N
SMILESCCSCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange to brown
Assay (GC)≥92.0%
FormLiquid
Refractive Index1.4685-1.4745 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only