Thermo Scientific Chemicals

Acetobromo-alpha-D-glucose, 97%, stab. with 2% calcium carbonate, Thermo Scientific Chemicals

Catalog number: L04151.09
10 g, Each
Thermo Scientific Chemicals

Acetobromo-alpha-D-glucose, 97%, stab. with 2% calcium carbonate, Thermo Scientific Chemicals

Catalog number: L04151.09
10 g, Each
Quantity
Catalog number: L04151.09
also known as L04151-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
572-09-8
IUPAC Name
[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Molecular Formula
C14H19BrO9
InChI Key
CYAYKKUWALRRPA-HAGGZJDPNA-N
SMILES
CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
Form
powder
Assay (Titration ex Bromide)
>96.0% (corrected for stabilser)
Optical Rotation
+193 ? 7? ( (c=3 in chloroform)
Appearance (Color)
White to pale cream or pale grey

Description

Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.

Solubility
Soluble in ether, chloroform, ethanol, acetone, dichloromethane, ether, ethyl, methanol, and tetrahydrofuran. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text