Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: L04156.06
5 g, Each
Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: L04156.06
5 g, Each
Quantity
Catalog number: L04156.06
also known as L04156-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
480-66-0
IUPAC Name
1-(2,4,6-trihydroxyphenyl)ethan-1-one
Molecular Formula
C8H8O4
InChI Key
XLEYFDVVXLMULC-UHFFFAOYSA-N
SMILES
CC(=O)C1=C(O)C=C(O)C=C1O
Form
Powder
Clarity
Clear (0.5% w/v solution in Ethanol)
Appearance (Color)
Cream to pale brown/orange
Water Content (Karl Fischer Titration)
<12.0%
Assay (HPLC)
>98.0%

Description

2',4',6'-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text