Thermo Scientific Chemicals

Chloro(methyl)diphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L04162.09
10 g, Each
Thermo Scientific Chemicals

Chloro(methyl)diphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L04162.09
10 g, Each
Quantity
Catalog number: L04162.09
also known as L04162-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144-79-6
IUPAC Name
chloro(methyl)diphenylsilane
Molecular Formula
C13H13ClSi
InChI Key
OJZNZOXALZKPEA-UHFFFAOYSA-N
SMILES
C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.5710-1.5760 @ 20?C
Form
Liquid

Description

Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text