Thermo Scientific Chemicals

1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals

Catalog number: L04181.18
50 g, Each
Thermo Scientific Chemicals

1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals

Catalog number: L04181.18
50 g, Each
Quantity
Catalog number: L04181.18
also known as L04181-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
513-36-0
IUPAC Name
1-chloro-2-methylpropane
Molecular Formula
C4H9Cl
InChI Key
QTBFPMKWQKYFLR-UHFFFAOYSA-N
SMILES
CC(C)CCl
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text