1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals
1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number L04181.30
also known as L04181-30
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals
Catalog numberL04181.30
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
CAS513-36-0
IUPAC Name1-chloro-2-methylpropane
Molecular FormulaC4H9Cl
InChI KeyQTBFPMKWQKYFLR-UHFFFAOYSA-N
SMILESCC(C)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Benito, J.; Guerrero, H.; Artigas, H.; López, M. C.; Lafuente, C. Refractive indices and static permittivities of systems containing n-hexane or n-heptane and isomeric chlorobutanes. J. Chem. Thermodyn. 2015, 86, 162-167.
  2. Montano, D.; Bölts, R.; Gmehling, J.; Artigas, H.; Lafuente, C. Calorimetric and acoustic study of binary mixtures containing an isomeric chlorobutane and butyl ethyl ether or methyl tert-butyl ether. J. Therm. Anal. Calorim. 2015, 122 (2), 851-861.