Thermo Scientific Chemicals

2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT, Thermo Scientific Chemicals

Catalog number: L04207.18
50 g, Each
Thermo Scientific Chemicals

2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT, Thermo Scientific Chemicals

Catalog number: L04207.18
50 g, Each
Quantity
Catalog number: L04207.18
also known as L04207-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
513-81-5
Assay (GC)
≥97.5%
Refractive Index
1.4365-1.4405 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.

Solubility
Miscible with chloroform. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text