Thermo Scientific Chemicals

4'-Methoxy-2'-nitroacetanilide, 98+%, Thermo Scientific Chemicals

Catalog number: L04218.14
25 g, Each
Thermo Scientific Chemicals

4'-Methoxy-2'-nitroacetanilide, 98+%, Thermo Scientific Chemicals

Catalog number: L04218.14
25 g, Each
Quantity
Catalog number: L04218.14
also known as L04218-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-81-3
IUPAC Name
N-(4-methoxy-2-nitrophenyl)acetamide
Molecular Formula
C9H10N2O4
InChI Key
QGEGALJODPBPGR-UHFFFAOYSA-N
SMILES
COC1=CC=C(NC(C)=O)C(=C1)[N+]([O-])=O
Appearance (Color)
Yellow to orange to brown
Melting Point (clear melt)
112.0-121.0?C
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text